Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-3-iodopropane 98% stabilised

Supplier: AMBEED, INC
Danger

Stabilization: Stabilised with copper chip

A391520-1G A391520-25G A391520-5G
77240-514EA 18 USD InStock
77240-514 77240-516 77240-518
1-Bromo-3-iodopropane 98% stabilised
1-Bromo-3-iodopropane
Precaución: for Research Use Only


Formula: C₃H₆BrI
MW: 248.88 g/mol
MDL Number: MFCD00040150
CAS Number: 22306-36-1
UN: 1993
ADR: 3 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty