Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(3-amino-4-hydroxyphenyl)sulfone 95%

Supplier: AMBEED, INC
Warning

A391746-10G A391746-1G A391746-25G A391746-5G
77278-608EA 27 USD InStock
77278-608 77278-610 77278-612 77278-614
Bis(3-amino-4-hydroxyphenyl)sulfone 95%
Bis(3-amino-4-hydroxyphenyl)sulfone
Precaución: for Research Use Only


Formula: C₁₂H₁₂N₂O₄S
MW: 280.30 g/mol
MDL Number: MFCD00025181
CAS Number: 7545-50-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty