Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxyethyl phenyl sulfide 97%

Supplier: AMBEED, INC
Warning

A392356-10G A392356-25G A392356-5G A392356-100G
77252-038EA 11 USD InStock
77252-038 77252-040 77252-042 77746-785
2-Hydroxyethyl phenyl sulfide 97%
2-Hydroxyethyl phenyl sulphide
Precaución: for Research Use Only


Formula: C₈H₁₀OS
MW: 154.23 g/mol
MDL Number: MFCD00002905
CAS Number: 699-12-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty