Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,6-Difluorosalicylic acid 95%

Supplier: AMBEED, INC
Warning

A392577-100MG A392577-1G A392577-250MG
77252-044EA 31 USD InStock
77252-044 77252-046 77252-048
5,6-Difluorosalicylic acid 95%
5,6-Difluorosalicylic acid
Precaución: for Research Use Only


Formula: C₇H₄F₂O₃
MW: 174.1017 g/mol
MDL Number: MFCD07368161
CAS Number: 749230-47-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty