Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(1H-1,2,4-Triazol-1-yl)benzaldehyde 98%

Supplier: AMBEED, INC
Warning

A395036-10G A395036-1G A395036-250MG A395036-5G
77264-464EA 264 USD InStock
77264-464 77264-466 77264-468 77264-470
2-(1H-1,2,4-Triazol-1-yl)benzaldehyde 98%
2-(1H-1,2,4-Triazol-1-yl)benzaldehyde
Precaución: for Research Use Only


Formula: C₉H₇N₃O
MDL Number: MFCD03617913
CAS Number: 138479-53-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty