Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Oxo-1,2-dihydro-3-quinolinecarboxylic acid 98%

Supplier: AMBEED, INC
Warning

A400246-100MG A400246-1G A400246-250MG A400246-5G
77278-710EA 15 USD InStock
77278-710 77278-712 77278-714 77278-716
2-Oxo-1,2-dihydro-3-quinolinecarboxylic acid 98%
2-Oxo-1,2-dihydro-3-quinolinecarboxylic acid
Precaución: for Research Use Only


Formula: C₁₀H₇NO₃
MDL Number: MFCD01550054
CAS Number: 2003-79-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty