Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(Hydroxymethyl)benzamide 98%

Supplier: AMBEED, INC
Warning

A400539-1000G A400539-100G A400539-25G A400539-500G
77252-128EA 136 USD InStock
77252-128 77252-130 77252-132 77252-134
N-(Hydroxymethyl)benzamide 98%
N-(Hydroxymethyl)benzamide
Precaución: for Research Use Only


Formula: C₈H₉NO₂
MW: 151.16 g/mol
MDL Number: MFCD00014415
CAS Number: 6282-02-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty