Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxydiphenylmethane 97%

Supplier: AMBEED, INC
Danger

A400708-1G A400708-5G
77291-136EA 30 USD InStock
77291-136 77291-138
4-Methoxydiphenylmethane 97%
4-Methoxydiphenylmethane
Precaución: for Research Use Only


Formula: C₁₄H₁₄O
MDL Number: MFCD00094072
CAS Number: 834-14-0
UN: 3082
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty