Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Oxobutyric acid 98%

Supplier: AMBEED, INC
Warning

A400901-100G A400901-1G A400901-5G
77264-510EA 720 USD InStock
77264-510 77264-512 77264-514
2-Oxobutyric acid 98%
2-Oxobutyric acid
Precaución: for Research Use Only


Formula: C₄H₆O₃
MDL Number: MFCD00004164
CAS Number: 600-18-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty