Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Cyclopropanecarboxamido)benzoic acid 98%

Supplier: AMBEED, INC
Danger

A401569-1G A401569-250MG A401569-5G
77291-140EA 50 USD InStock
77291-140 77291-142 77291-144
4-(Cyclopropanecarboxamido)benzoic acid 98%
4-(Cyclopropanecarboxamido)benzoic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₁NO₃
MDL Number: MFCD02602219
CAS Number: 23745-26-8
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty