Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sbc-115076 98+%

Supplier: AMBEED, INC
Warning

A402455-10MG A402455-1MG A402455-25MG A402455-5MG
77240-636EA 64 USD InStock
77240-636 77240-638 77240-640 77240-642
Sbc-115076 98+%
Sbc-115076
Precaución: for Research Use Only


Formula: C₃₁H₃₃N₃O₅
MW: 527.61 g/mol
MDL Number: MFCD03296540
CAS Number: 489415-96-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty