Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Guanidinopropionic acid 98%

Supplier: AMBEED, INC
Warning

A403091-100G A403091-10G A403091-1G A403091-25G A403091-500G A403091-5G
77291-164EA 44 USD InStock
77291-164 77291-166 77291-168 77291-170 77291-172 77291-174
3-Guanidinopropionic acid 98%
3-Guanidinopropionic acid


Formula: C₄H₉N₃O₂
MDL Number: MFCD00045939
CAS Number: 353-09-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty