Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(16aS)-1,16-Bis(diphenylphosphino)-6,7,8,9,10,11-hexahydrodibenzo[b,d][1,6]dioxacyclododecine 98%

Supplier: AMBEED, INC
Warning

A403133-100MG A403133-1G A403133-250MG
77264-532EA 110 USD InStock
77264-532 77264-534 77264-536
(16aS)-1,16-Bis(diphenylphosphino)-6,7,8,9,10,11-hexahydrodibenzo[b,d][1,6]dioxacyclododecine 98%
(16aS)-1,16-Bis(diphenylphosphino)-6,7,8,9,10,11-hexahydrodibenzo[b,d][1,6]dioxacyclododecine
Precaución: for Research Use Only


Formula: C₄₂H₃₈O₂P₂
CAS Number: 486429-96-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty