Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-3-nitrophenyl methyl sulfone 98%

Supplier: AMBEED, INC
Warning

A403148-1G A403148-25G A403148-5G
77291-176EA 10 USD InStock
77291-176 77291-178 77291-180
4-Fluoro-3-nitrophenyl methyl sulfone 98%
4-Fluoro-3-nitrophenyl methyl sulfone
Precaución: for Research Use Only


Formula: C₇H₆FNO₄S
MDL Number: MFCD00025066
CAS Number: 453-72-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty