Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-N-isopropylbenzamide 97%

Supplier: AMBEED, INC
Warning

A404002-100MG A404002-1G A404002-250MG A404002-5G
77278-780EA 14 USD InStock
77278-780 77278-782 77278-784 77278-786
3-Amino-N-isopropylbenzamide 97%
3-Amino-N-isopropylbenzamide
Precaución: for Research Use Only


Formula: C₁₀H₁₄N₂O
MDL Number: MFCD09045316
CAS Number: 81882-62-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty