Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-[4-(Bromomethyl)phenyl]propionic acid 95%

Supplier: AMBEED, INC
Danger

A404623-100G A404623-10G A404623-25G A404623-500G A404623-5G
77278-788EA 174.07 USD InStock
77278-788 77278-790 77278-792 77278-794 77278-796 77278-798
2-[4-(Bromomethyl)phenyl]propionic acid 95%
2-[4-(Bromomethyl)phenyl]propionic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₁BrO₂
MDL Number: MFCD02093445
CAS Number: 111128-12-2
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty