Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Z)-2-(3,4-Dimethoxyphenyl)-3-(5-(4-hydroxypiperidin-1-yl)thiophen-2-yl)acrylonitrile 98+%

Supplier: AMBEED, INC
Warning

A407213-1G A407213-1MG A407213-5MG
77240-698EA 440 USD InStock
77240-698 77240-700 77240-702
(Z)-2-(3,4-Dimethoxyphenyl)-3-(5-(4-hydroxypiperidin-1-yl)thiophen-2-yl)acrylonitrile 98+%
(Z)-2-(3,4-Dimethoxyphenyl)-3-(5-(4-hydroxypiperidin-1-yl)thiophen-2-yl)acrylonitrile
Precaución: for Research Use Only


Formula: C₂₀H₂₂N₂O₃S
MDL Number: MFCD28900680
CAS Number: 912287-56-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty