Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-2-chloro-5-fluorotoluene 98%

Supplier: AMBEED, INC
Warning

A407508-10G A407508-1G A407508-25G A407508-5G
77291-246EA 36 USD InStock
77291-246 77291-248 77291-250 77291-252
4-Bromo-2-chloro-5-fluorotoluene 98%
4-Bromo-2-chloro-5-fluorotoluene
Precaución: for Research Use Only


Formula: C₇H₅BrClF
MDL Number: MFCD00236592
CAS Number: 93765-83-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty