Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Butoxybenzoic acid 98%

Supplier: AMBEED, INC
Warning

A407665-1G A407665-25G A407665-5G
77291-254EA 6 USD InStock
77291-254 77291-256 77291-258
4-Butoxybenzoic acid 98%
4-Butoxybenzoic acid


Formula: C₁₁H₁₄O₃
MDL Number: MFCD00002546
CAS Number: 1498-96-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty