Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Diamino-6-dimethylamino-1,3,5-triazine 98%

Supplier: AMBEED, INC
Warning

A408251-1G A408251-250MG A408251-5G A408251-100MG
77252-196EA 31 USD InStock
77252-196 77252-198 77252-200 77746-823
2,4-Diamino-6-dimethylamino-1,3,5-triazine 98%
2,4-Diamino-6-dimethylamino-1,3,5-triazine
Precaución: for Research Use Only


Formula: C₅H₁₀N₆
MDL Number: MFCD00191338
CAS Number: 1985-46-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty