Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-[Bis(3,5-trifluoromethylphenylphosphino)-3,6-dimethoxy]- 2',6'-di-i-propoxy-1,1'-biphenyl 98%

Supplier: AMBEED, INC
Warning

A409134-250MG A409134-50MG A409134-100MG
77252-204EA 90 USD InStock
77252-204 77746-826 77746-825
2-[Bis(3,5-trifluoromethylphenylphosphino)-3,6-dimethoxy]- 2',6'-di-i-propoxy-1,1'-biphenyl 98%
2-[Bis(3,5-trifluoromethylphenylphosphino)-3,6-dimethoxy]- 2',6'-di-i-propoxy-1,1'-biphenyl
Precaución: for Research Use Only


Formula: C₃₆H₃₁F₁₂O₄P
MDL Number: MFCD29905025
CAS Number: 1810068-31-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty