Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-3-cyano-4-methoxypyridine 98%

Supplier: AMBEED, INC
Danger

A410275-1G A410275-250MG A410275-25G A410275-5G
77264-582EA 15 USD InStock
77264-582 77264-584 77264-586 77264-588
2-Chloro-3-cyano-4-methoxypyridine 98%
2-Chloro-3-cyano-4-methoxypyridine
Precaución: for Research Use Only


Formula: C₇H₅ClN₂O
MW: 168.5804 g/mol
MDL Number: MFCD00975444
CAS Number: 98645-43-3
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty