Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4-Dimethylpiperidine 98%

Supplier: AMBEED, INC
Danger

A410897-100MG A410897-1G A410897-250MG A410897-5G
77291-300EA 19 USD InStock
77291-300 77291-302 77291-304 77291-306
4,4-Dimethylpiperidine 98%
4,4-Dimethylpiperidine
Precaución: for Research Use Only


Formula: C₇H₁₅N
MDL Number: MFCD04037433
CAS Number: 4045-30-1
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty