Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-3,3'-Bis3,5-bis(trifluoromethyl)phenyl)-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2,2'-naphthol 98%

Supplier: AMBEED, INC
Warning

A411408-100MG A411408-1G A411408-250MG A411408-5G
77252-216EA 16 USD InStock
77252-216 77252-218 77252-220 77746-832
(S)-3,3'-Bis3,5-bis(trifluoromethyl)phenyl)-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2,2'-naphthol 98%
(S)-3,3'-Bis3,5-bis(trifluoromethyl)phenyl)-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2,2'-naphthol
Precaución: for Research Use Only


Formula: C₃₆H₂₆F₁₂O₂
MDL Number: MFCD31560041
CAS Number: 1228600-99-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty