Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bromo-PEG2-bromide 98%

Supplier: AMBEED, INC
Warning

A411817-1G A411817-250MG A411817-5G A411817-100MG
77264-590EA 72 USD InStock
77264-590 77264-592 77264-594 77748-830
Bromo-PEG2-bromide 98%
Bromo-PEG2-bromide
Precaución: for Research Use Only


Formula: C₆H₁₂Br₂O₂
MDL Number: MFCD00466590
CAS Number: 31255-10-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty