Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Bis((2S,5S)-2,5-di-i-propylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate 98%

Supplier: AMBEED, INC
Warning

A412533-100MG A412533-1G A412533-250MG
77252-248EA 58 USD InStock
77252-248 77252-250 77252-252
1,2-Bis((2S,5S)-2,5-di-i-propylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate 98%
1,2-Bis((2S,5S)-2,5-di-i-propylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate
Precaución: for Research Use Only


Formula: C₃₀H₅₄BF₄P₂Rh
MDL Number: MFCD17018835
CAS Number: 213343-68-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty