Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Mivebresib 97%

Supplier: AMBEED, INC
Warning

A414117-10MG A414117-1MG A414117-25MG A414117-2MG A414117-5MG A414117-50MG
77264-626EA 67 USD InStock
77264-626 77264-628 77264-630 77264-632 77264-634 77748-837
Mivebresib 97%
Mivebresib
Precaución: for Research Use Only


Formula: C₂₂H₁₉F₂N₃O₄S
MDL Number: MFCD30377200
CAS Number: 1445993-26-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty