Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Cyclopentyl-2-((4-(4-methylpiperazin-1-yl)phenyl)amino)-7-oxo-7,8-dihydropyrido[2,3-d]pyrimidine-6-carbonitrile 98%

Supplier: AMBEED, INC
Warning

A414519-10MG A414519-1MG A414519-2MG A414519-50MG A414519-5MG
77240-764EA 61 USD InStock
77240-764 77240-766 77240-768 77240-770 77240-772
8-Cyclopentyl-2-((4-(4-methylpiperazin-1-yl)phenyl)amino)-7-oxo-7,8-dihydropyrido[2,3-d]pyrimidine-6-carbonitrile 98%
8-Cyclopentyl-2-((4-(4-methylpiperazin-1-yl)phenyl)amino)-7-oxo-7,8-dihydropyrido[2,3-d]pyrimidine-6-carbonitrile
Precaución: for Research Use Only


Formula: C₂₄H₂₇N₇O
MW: 429.52 g/mol
MDL Number: MFCD28411414
CAS Number: 1357470-29-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty