Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-3-Methoxycinnamaldehyde 95%

Supplier: AMBEED, INC
Warning

A414906-100MG A414906-1MG A414906-25MG A414906-5G A414906-250MG
77291-342EA 20 USD InStock
77291-342 77291-344 77291-346 77748-839 77748-838
4-Hydroxy-3-Methoxycinnamaldehyde 95%
4-Hydroxy-3-Methoxycinnamaldehyde
Precaución: for Research Use Only


Formula: C₁₀H₁₀O₃
MW: 178.18 g/mol
MDL Number: MFCD00075811
CAS Number: 458-36-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty