Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

CCG-203971 97%

Supplier: AMBEED, INC
Warning

A415352-100MG A415352-10MG A415352-1MG A415352-25MG A415352-50MG
77264-650EA 139 USD InStock
77264-650 77264-652 77264-654 77264-656 77264-658
CCG-203971 97%
CCG-203971
Precaución: for Research Use Only


Formula: C₂₃H₂₁ClN₂O₃
MDL Number: MFCD28166491
CAS Number: 1443437-74-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty