Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-2,2-difluoro-1,3-benzodioxole 96%

Supplier: AMBEED, INC
Danger

A416417-250MG A416417-5G
77291-360EA 39 USD InStock
77291-360 77291-362
4-Amino-2,2-difluoro-1,3-benzodioxole 96%
4-Amino-2,2-difluoro-1,3-benzodioxole
Precaución: for Research Use Only


Formula: C₇H₅F₂NO₂
MW: 173.12
MDL Number: MFCD00792416
CAS Number: 106876-54-4
UN: 2810
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty