Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dimethylimidazole 96%

Supplier: AMBEED, INC
Danger

A416867-10G A416867-25G A416867-5G
77252-302EA 6 USD InStock
77252-302 77252-304 77252-306
2,4-Dimethylimidazole 96%
2,4-Dimethylimidazole
Precaución: for Research Use Only


Formula: C₅H₈N₂
MW: 96.13
Boiling Pt: 266
Melting Pt: 92
MDL Number: MFCD00022365
CAS Number: 930-62-1
UN: 3263
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty