Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-3-methylbutanamide 97%

Supplier: AMBEED, INC
Warning

A417247-100MG A417247-250MG
77264-666EA 20 USD InStock
77264-666 77264-668
3-Amino-3-methylbutanamide 97%
3-Amino-3-methylbutanamide
Precaución: for Research Use Only


Formula: C₅H₁₂N₂O
MDL Number: MFCD19205344
CAS Number: 173337-04-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty