Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-4-Fluoro-β-nitrostyrene 98% mix stabilized

Supplier: AMBEED, INC
Danger

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A417856-100G A417856-25G A417856-5G
77291-366EA 261 USD InStock
77291-366 77291-368 77291-370
trans-4-Fluoro-β-nitrostyrene 98% mix stabilized
trans-4-Fluoro-β-nitrostyrene
Precaución: for Research Use Only


Formula: C₈H₆FNO₂
MDL Number: MFCD00024824
CAS Number: 706-08-1
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty