Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-methoxypyrazine 98%

Supplier: AMBEED, INC
Warning

A422156-100G A422156-100MG A422156-10G A422156-1G A422156-25G
77264-722EA 167 USD InStock
77264-722 77264-724 77264-726 77264-728 77264-730 77264-732
2-Chloro-6-methoxypyrazine 98%
2-Chloro-6-methoxypyrazine
Precaución: for Research Use Only


Formula: C₅H₅ClN₂O
MDL Number: MFCD06410937
CAS Number: 33332-30-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty