Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-7-chloro-2-oxoheptanoate 97%

Supplier: AMBEED, INC
Warning

A423288-10G A423288-1G A423288-25G A423288-500G A423288-5G A423288-100G
77252-358EA 8 USD InStock
77252-358 77252-360 77252-362 77252-364 77252-366 77746-853
Ethyl-7-chloro-2-oxoheptanoate 97%
Ethyl-7-chloro-2-oxoheptanoate
Precaución: for Research Use Only


Formula: C₉H₁₅ClO₃
MDL Number: MFCD07698675
CAS Number: 78834-75-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty