Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium 2-cyanoacetate 97%

Supplier: AMBEED, INC
Warning

A424879-100G A424879-1G A424879-25G A424879-5G
77252-372EA 231 USD InStock
77252-372 77252-374 77252-376 77252-378
Potassium 2-cyanoacetate 97%
Potassium 2-cyanoacetate
Precaución: for Research Use Only


Formula: C₃H₂KNO₂
MDL Number: MFCD11498991
CAS Number: 7062-95-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty