Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-5-isopropyl-2-(perfluorophenyl)-2,5,6,8-tetrahydro-[1,2,4]triazolo[3,4-c][1,4]oxazin-4-ium tetrafluoroborate 98%

Supplier: AMBEED, INC
Danger

A427278-100MG A427278-250MG
77252-416EA 49 USD InStock
77252-416 77252-418
(S)-5-isopropyl-2-(perfluorophenyl)-2,5,6,8-tetrahydro-[1,2,4]triazolo[3,4-c][1,4]oxazin-4-ium tetrafluoroborate 98%
(S)-5-isopropyl-2-(perfluorophenyl)-2,5,6,8-tetrahydro-[1,2,4]triazolo[3,4-c][1,4]oxazin-4-ium tetrafluoroborate
Precaución: for Research Use Only


Formula: C₁₄H₁₃BF₉N₃O
MW: 421.06 g/mol
MDL Number: MFCD32655222
CAS Number: 1431323-18-0
UN: 1759
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty