Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Nitrostyrene 95%

Supplier: AMBEED, INC
Warning

A429202-100MG A429202-10G A429202-1G A429202-250MG A429202-5G
77291-494EA 22 USD InStock
77291-494 77291-496 77291-498 77291-500 77291-502
3-Nitrostyrene 95%
3-Nitrostyrene
Precaución: for Research Use Only


Formula: C₈H₇NO₂
MDL Number: MFCD00007276
CAS Number: 586-39-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty