Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Bis(4-chlorophenyl)acetonitrile 97%

Supplier: AMBEED, INC
Warning

A430424-100MG A430424-1G A430424-250MG
77240-870EA 29 USD InStock
77240-870 77240-872 77240-874
2,2-Bis(4-chlorophenyl)acetonitrile 97%
2,2-Bis(4-chlorophenyl)acetonitrile
Precaución: for Research Use Only


Formula: C₁₄H₉Cl₂N
MDL Number: MFCD19303617
CAS Number: 20968-04-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty