Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

98%

Supplier: AMBEED, INC
Warning

A431215-10G A431215-1G A431215-250MG A431215-5G
77279-048EA 418 USD InStock
77279-048 77279-050 77279-052 77279-054
98%
3,4-Dimethoxybenzamide
Precaución: for Research Use Only


Formula: C₉H₁₁NO₃
MDL Number: MFCD00017128
CAS Number: 1521-41-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty