Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-(4-formylphenyl)benzoate 98%

Supplier: AMBEED, INC
Danger

A431370-250MG A431370-1G
77291-530EA 15 USD InStock
77291-530 77748-872
Methyl 4-(4-formylphenyl)benzoate 98%
Methyl 4-(4-formylphenyl)benzoate
Precaución: for Research Use Only


Formula: C₁₅H₁₂O₃
MW: 240.25 g/mol
MDL Number: MFCD04039123
CAS Number: 70916-89-1
UN: 3077
ADR: 9 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty