Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methoxy-p-toluic acid 98%

Supplier: AMBEED, INC
Warning

A432270-100G A432270-10G A432270-1G A432270-25G A432270-5G
77264-872EA 294 USD InStock
77264-872 77264-874 77264-876 77264-878 77748-874
2-Methoxy-p-toluic acid 98%
2-Methoxy-p-toluic acid
Precaución: for Research Use Only


Formula: C₉H₁₀O₃
MDL Number: MFCD00020380
CAS Number: 704-45-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty