Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4'-Bis(4-aminophenoxy)biphenyl 98%

Supplier: AMBEED, INC
Warning

A432321-100G A432321-25G A432321-5G A432321-500G
77291-532EA 62 USD InStock
77291-532 77291-534 77291-536 77748-875
4,4'-Bis(4-aminophenoxy)biphenyl 98%
4,4'-Bis(4-aminophenoxy)biphenyl
Precaución: for Research Use Only


Formula: C₂₄H₂₀N₂O₂
MDL Number: MFCD00039151
CAS Number: 13080-85-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty