Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(+)-Bis-(1,2-diphenylphosphino)propane 98%

Supplier: AMBEED, INC
Warning

A434030-100MG A434030-1G A434030-250MG
77291-556EA 66 USD InStock
77291-556 77291-558 77291-560
(R)-(+)-Bis-(1,2-diphenylphosphino)propane 98%
(R)-(+)-Bis-(1,2-diphenylphosphino)propane
Precaución: for Research Use Only


Formula: C₂₇H₂₆P₂
MW: 412.453 g/mol
MDL Number: MFCD00012309
CAS Number: 67884-32-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty