Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(E)-N-(3-([3,3'-bithiophen]-5-ylmethoxy)benzyl)-N-ethyl-6,6-dimethylhept-2-en-4-yn-1-amine 98%

Supplier: AMBEED, INC
Warning

A436262-25MG A436262-50MG
77240-930EA 172 USD InStock
77240-930 77240-932
(E)-N-(3-([3,3'-bithiophen]-5-ylmethoxy)benzyl)-N-ethyl-6,6-dimethylhept-2-en-4-yn-1-amine 98%
(E)-N-(3-([3,3'-bithiophen]-5-ylmethoxy)benzyl)-N-ethyl-6,6-dimethylhept-2-en-4-yn-1-amine
Precaución: for Research Use Only


Formula: C₂₇H₃₁NOS₂
MDL Number: MFCD00898838
CAS Number: 131060-14-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty