Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Bromophenoxy)trimethylsilane 98%

Supplier: AMBEED, INC
Warning

A436393-100G A436393-1G A436393-25G A436393-5G
77264-932EA 140 USD InStock
77264-932 77264-934 77264-936 77264-938
(4-Bromophenoxy)trimethylsilane 98%
(4-Bromophenoxy)trimethylsilane


Formula: C₉H₁₃BrOSi
MDL Number: MFCD00040713
CAS Number: 17878-44-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty