Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-1-phenyl-2-propanol 98%

Supplier: AMBEED, INC
Warning

A437483-1000G A437483-100G A437483-25G A437483-500G
77264-940EA 53 USD InStock
77264-940 77264-942 77264-944 77264-946
2-Methyl-1-phenyl-2-propanol 98%
2-Methyl-1-phenyl-2-propanol
Precaución: for Research Use Only


Formula: C₆H₅CH₂C(CH₃)(OH)CH₃
MDL Number: MFCD00004465
CAS Number: 100-86-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty