Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Coumalic acid 97%

Supplier: AMBEED, INC
Warning

A437681-100G A437681-10G A437681-1G A437681-250MG A437681-25G A437681-500G
77279-090EA 190 USD InStock
77279-090 77279-092 77279-094 77279-096 77279-098 77279-100
Coumalic acid 97%
Coumalic acid
Precaución: for Research Use Only


Formula: C₆H₄O₄
MDL Number: MFCD00006644
CAS Number: 500-05-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty