Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Bromoethyl)pyridine hydrobromide 98%

Supplier: AMBEED, INC
Danger

A438513-1G A438513-250MG A438513-25G A438513-5G
77240-942EA 37 USD InStock
77240-942 77240-944 77240-946 77240-948
2-(2-Bromoethyl)pyridine hydrobromide 98%
2-(2-Bromoethyl)pyridine hydrobromide
Precaución: for Research Use Only


Formula: C₇H₉Br₂N
MDL Number: MFCD08274972
CAS Number: 72996-65-7
UN: 1759
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty